EMAIL THIS PAGE TO A FRIEND
Marine Biotechnology
Alginate Microencapsulation for Oral Immunisation of Finfish: Release Characteristics, Ex Vivo Intestinal Uptake and In Vivo Administration in Atlantic Salmon, Salmo salar L.
PMID 26410294