EMAIL THIS PAGE TO A FRIEND
Toxicology Letters
Gestational N-hexane inhalation alters the expression of genes related to ovarian hormone production and DNA methylation states in adult female F1 rat offspring.
PMID 26410608