EMAIL THIS PAGE TO A FRIEND
Chirality
A Solid Phase Vibrational Circular Dichroism Study of Polypeptide-Surfactant Interaction.
PMID 26413930
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4762
Myristyltrimethylammonium bromide, ≥99%
C17H38BrN