EMAIL THIS PAGE TO A FRIEND
Oncotarget
Acute MUS81 depletion leads to replication fork slowing and a constitutive DNA damage response.
PMID 26415217
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU063801 MISSION® esiRNA, esiRNA targeting mouse Eme1 (esiRNA1)