EMAIL THIS PAGE TO A FRIEND
Toxicology and Applied Pharmacology
Compound- and sex-specific effects on programming of energy and immune homeostasis in adult C57BL/6JxFVB mice after perinatal TCDD and PCB 153.
PMID 26415833