EMAIL THIS PAGE TO A FRIEND
Transplant immunology
Hypodermin A, a potential agent for prevention of allogeneic acute rejection.
PMID 26416179
Abstract
Immunosuppressive agents play an important role in the success of organ transplantation, however the chronic toxicity of these agents is a major issue over the long-term. Hypodermin A (HA) is an enzyme secreted by the larvae of Hypoderma lineatum (Diptera: Oestridae), and has been implicated in immunosuppression in cattle. Malassagne et al. have demonstrated that HA can degrade the C3 protein, and could be used to prevent hyperacute xenogeneic rejection. We found that overexpression of HA in RAW264.7 cells induced significant secretion of prostaglandin E2 (PGE2), which mediates a variety of innate and adaptive immune responses through four E-type prostanoid (EP) receptor subtypes (EP1-4). PGE2 is useful in the management of allogeneic acute rejection. In addition, we found that induction of PGE2 expression downregulates the expression of interferon (IFN)-γ and interleukin (IL)-2, and promotes the secretion of IL-10 in vitro through the EP4 receptor. It was previously shown that activation of IL-2 and IFN-γ is involved in allograft acute rejection. IL-10 is known to prevent inflammation, and can improve allograft survival rates. We concluded that besides preventing hyperacute xenogeneic rejection, HA might also be a potential therapeutic candidate for ameliorating acute rejection during allotransplantation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N194
NS-398, ≥98% (HPLC), solid
C13H18N2O5S