EMAIL THIS PAGE TO A FRIEND
Journal of food science
Structure and Antioxidant Activities of Proanthocyanidins from Elephant Apple (Dillenia indica Linn.).
PMID 26416622
Abstract
Proanthocyanidins were isolated and purified from fruits of elephant apple (Dillenia indica Linn.) and their structural and bioactive properties were examined. Bate-Smith alcoholysis, FTIR, and (13) C NMR spectra revealed that elephant apple proanthocyanidins (EAPs) contained a dominant amount of B-type procyanidins (PC) with a minor amount of B-type prodelphinidins (PD) but no A-type interflavan linkage. (13) C NMR spectrum indicated that the cis isomer was dominant in EAPs. The electron spray ionization and matrix-assisted laser desorption ionization time of flight mass spectra of EAPs showed the clear ion peaks corresponding to B-type PC dimer to B-type PD with degree of polymerization of 11. EAPs had strong antioxidant activity, which was evidenced by the high oxygen radical scavenging capacity at 1.06 × 10(4) μmol TE/g and ferric reducing antioxidant power of 2320 μmol Fe(II)/g. The results suggest that EAPs could be extracted to be used as promising functional food materials. In this study, the elephant apple proanthocyanidins (EAPs) with a yield of 0.23% were identified for the first time as dominant B-type poly(catechin/epicatechin) but no A-type interflavan linkage. EAPs had higher ORAC and FRAP values compared to commercial grape seed proanthocyanidins, suggesting that EAPs may be used as promising functional food materials.