EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
A Fi??i??rster Resonance Energy Transfer (FRET)-based System Provides Insight into the Ordered Assembly of Yeast Septin Hetero-octamers.
PMID 26416886