EMAIL THIS PAGE TO A FRIEND
Journal of diabetes investigation
Fructose induces glucose-dependent insulinotropic polypeptide, glucagon-like peptide-1 and insulin secretion: Role of adenosine triphosphate-sensitive K(+) channels.
PMID 26417408
Abstract
Adenosine triphosphate-sensitive K(+) (KATP) channels play an essential role in glucose-induced insulin secretion from pancreatic β-cells. It was recently reported that the KATP channel is also found in the enteroendocrine K-cells and L-cells that secrete glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1), respectively. In the present study, we investigated the involvement of the KATP channel in fructose-induced GIP, GLP-1 and insulin secretion in mice. Fructose stimulated GIP secretion, but pretreatment with diazoxide, a KATP channel activator, did not affect fructose-induced GIP secretion under streptozotocin-induced hyperglycemic conditions. Fructose significantly stimulated insulin secretion in Kir6.2 (+/+) mice, but not in mice lacking KATP channels (Kir6.2 (-/-) ), and fructose stimulated GLP-1 secretion in both Kir6.2 (+/+) mice and Kir6.2 (-/-) mice under the normoglycemic condition. In addition, diazoxide completely blocked fructose-induced insulin secretion in Kir6.2 (+/+) mice and in MIN6-K8 β-cells. These results show that fructose-induced GIP and GLP-1 secretion is KATP channel-independent and that fructose-induced insulin secretion is KATP channel-dependent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9035
Diazoxide
C8H7ClN2O2S