EMAIL THIS PAGE TO A FRIEND
Cancer Biology & Therapy
Hodgkin lymphoma cell lines bind to platelets. Incubation with platelets induces CD15 and P-selectin dependent adhesion of the cell lines to Human Umbilical Vein Endothelial cells (HUVEC).
PMID 26418972