EMAIL THIS PAGE TO A FRIEND
Biomicrofluidics
Microfluidic sampling system for tissue analytics.
PMID 26421088
Abstract
We have developed a microfluidics based sampling system for tissue analytics. The proof-of-concept of the sampling system was demonstrated by extracting lipid samples from tissue biopsies. The sample collection system consists of a disposable silicon based multiport microneedle integrated with polymer microfluidics. The polymethyl methacrylate polymer microfluidic chip has a 10 μl sample reservoir and actuation membranes for liquid pumping. A special automated robotic system was developed to control the positioning of the needle and the sampling procedure on preselected spots on the tissue. Real breast cancer tissue samples were used to test the feasibility of the sampling system. We successfully measured indicative cancer biomarkers from the tissue surface. Phosphatidylcholine and phosphoethanolamine were extracted from the tissue membrane with methyl tert-butyl ether solvent and detected by mass spectrometry. In the future, this tool could be used in characterization of preoperative biopsies and tumour tissues removed during surgery.