EMAIL THIS PAGE TO A FRIEND
Cytotherapy
Quantitative activation suppression assay to evaluate human bone marrow-derived mesenchymal stromal cell potency.
PMID 26422657
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
460036
5-Norbornene-2-endo,3-exo-dicarboxylic acid, 97%
C9H10O4