EMAIL THIS PAGE TO A FRIEND
Sensors (Basel, Switzerland)
Optimisation and Characterisation of Anti-Fouling Ternary SAM Layers for Impedance-Based Aptasensors.
PMID 26426017
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W349518
1,6-Hexanedithiol, ≥97%, FG
C6H14S2
H12005
1,6-Hexanedithiol, 96%
C6H14S2
725382
1,6-Hexanedithiol, 99.5%
C6H14S2
451088
6-Mercapto-1-hexanol, 97%
C6H14OS
725226
6-Mercapto-1-hexanol, 99%
C6H14OS
455989
Potassium hexacyanoferrate(II) trihydrate, ≥99.95% trace metals basis
C6FeK4N6 · 3H2O
P3289
Potassium hexacyanoferrate(II) trihydrate, ACS reagent, 98.5-102.0%
C6FeK4N6 · 3H2O
P9387
Potassium hexacyanoferrate(II) trihydrate, ReagentPlus®, ≥98.5%
C6FeK4N6 · 3H2O
60279
Potassium hexacyanoferrate(II) trihydrate, BioUltra, ≥99.5% (RT)
C6FeK4N6 · 3H2O
31254
Potassium hexacyanoferrate(II) trihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
C6FeK4N6 · 3H2O
12639
Potassium hexacyanoferrate(II) trihydrate, purum, ≥99%
C6FeK4N6 · 3H2O
455946
Potassium hexacyanoferrate(III), 99.98% trace metals basis
C6FeK3N6
393517
Potassium hexacyanoferrate(III), ACS reagent, ≥99%, powder, <10 μm
C6FeK3N6
244023
Potassium hexacyanoferrate(III), ACS reagent, ≥99.0%
C6FeK3N6
60299
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
C6FeK3N6
31253
Potassium hexacyanoferrate(III), puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
C6FeK3N6
P8131
Potassium hexacyanoferrate(III), ReagentPlus®, ~99%
C6FeK3N6
12643
Potassium hexacyanoferrate(III), purum, ≥99% (iodometric)
C6FeK3N6
P9458
Potassium sulfate, BioXtra, ≥99.0%
K2O4S
RES0797P-A7
Potassium sulfate, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
K2O4S
P0772
Potassium sulfate, ReagentPlus®, ≥99.0%
K2O4S
P8541
Potassium sulfate, plant cell culture tested
K2O4S
204129
Potassium sulfate, 99.99% trace metals basis
K2O4S
221325
Potassium sulfate, ACS reagent, ≥99.0%, powder or crystals
K2O4S
223492
Potassium sulfate, ACS reagent, ≥99.0%, powder
K2O4S
60528
Potassium sulfate, BioUltra, ≥99.0% (AT)
K2O4S
31270
Potassium sulfate, puriss. p.a., ACS reagent, reag. ISO, ≥99%
K2O4S
12658
Potassium sulfate, meets analytical specification of Ph. Eur., 99-101%
K2O4S