EMAIL THIS PAGE TO A FRIEND
ACS Nano
Controlled DNA Patterning by Chemical Lift-Off Lithography: Matrix Matters.
PMID 26426585
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674249
11-Mercapto-1-undecanol, 99%
C11H24OS
447528
11-Mercapto-1-undecanol, 97%
C11H24OS