EMAIL THIS PAGE TO A FRIEND
ACS Nano
Holey Graphene as a Weed Barrier for Molecules.
PMID 26426746
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
659452
1-Adamantanethiol, 95%
C10H16S
719234
1-Adamantanethiol, 99% (GC)
C10H16S