EMAIL THIS PAGE TO A FRIEND
Clinical laboratory
Ciculating miRNA-21 as a Biomarker Predicts Polycystic Ovary Syndrome (PCOS) in Patients.
PMID 26427146
Abstract
Polycystic ovary syndrome (PCOS) is characterized by hyperandrogenism, hyperinsulinemia, and infertility. In PCOS, abnormal regulation of relevant genes is required for follicular development. By binding to the 3' untranslated region (3'URT), microRNAs (miRNAs) are widely involved in posttranscriptional gene regulation. However, few studies have been conducted on circulating miRNA expression in PCOS. This study aims to describe altered expression of circulating miR-21 in PCOS. The expression of serum miRNAs of PCOS patients were explored using the TaqMan Low Density Array followed by individual quantitative reverse transcription polymerase chain reaction assays. The protein level of LATS1 was determined using Western blot. To validate whether miR-21 targeted LATS1, the luciferase assay was applied. In comparison with normal subjects, the circulating level of miRNA-21 was significantly enhanced in PCOS patients. In PCOS patients, the expression levels of MST1/2, LATS1/2, TAZ were much lower than the control subjects. Luciferase reporter assay revealed that LATS1 was a downstream target of miR-21. In comparison with normal subjects, serum miR-21 is obviously increased in PCOS patients. Through targeting LATS1, miR-21 could prompt PCOS progression and could act as a novel non-invasive biomarker for diagnosis of PCOS.