EMAIL THIS PAGE TO A FRIEND
Carbohydrate Polymers
Immunomodulatory activity on macrophage of a purified polysaccharide extracted from Laminaria japonica.
PMID 26428101
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47829
D-(+) Glucose, analytical standard
C6H12O6
G0750
D-(+)-Galactose, ≥99%
C6H12O6
G0625
D-(+)-Galactose, ≥98%
C6H12O6
G6404
D-(+)-Galactose, BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), ≥99%
C6H12O6
G5388
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H12O6
15522
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
C6H12O6
G7528
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
C6H12O6
G8270
D-(+)-Glucose, ≥99.5% (GC)
C6H12O6
G5767
D-(+)-Glucose, ACS reagent
C6H12O6
G7021
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
C6H12O6
G5400
D-(+)-Glucose, ≥99% (GC)
C6H12O6
G6152
D-(+)-Glucose, suitable for mouse embryo, ≥99.5% (GC)
C6H12O6
G5146
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
C6H12O6
G7520
D-(+)-Glucose, plant cell culture tested, BioReagent
C6H12O6
49139
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
16325
D-(+)-Glucose, meets analytical specification of Ph. Eur., BP, anhydrous
C6H12O6
608203
D-(+)-Glucose, 99.9 atom % 16O, 99.9 atom % 12C
C6H12O6
47249
D-(+)-Glucose, analytical standard
C6H12O6
ARK2181
D-(+)-Mannose, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C6H12O6
M2069
D-(+)-Mannose, from wood, ≥99%
C6H12O6
M8574
D-(+)-Mannose, synthetic, ≥99%
C6H12O6
M6020
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
C6H12O6
63579
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
M4625
D-(+)-Mannose
C6H12O6
W360600
D-(+)-Xylose, ≥99%
C5H10O5
X1500
D-(+)-Xylose, ≥99%
C5H10O5
X3877
D-(+)-Xylose, BioXtra, ≥99%
C5H10O5
95729
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C5H10O5
47253
D-(+)-Xylose, analytical standard
C5H10O5
D9434
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
C6H12O6