EMAIL THIS PAGE TO A FRIEND
Carbohydrate Polymers
Facile approach to synthesize chitosan based composite--Characterization and cadmium(II) ion adsorption studies.
PMID 26428168
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282979
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
C7H12O2
20911
Cadmium nitrate tetrahydrate, purum p.a., ≥99.0% (T)
CdN2O6 · 4H2O
229520
Cadmium nitrate tetrahydrate, 99.997% trace metals basis
CdN2O6 · 4H2O
642045
Cadmium nitrate tetrahydrate, 98%
CdN2O6 · 4H2O
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
435570
Hydrochloric acid, reagent grade, 37%
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
84434
Hydrochloric acid solution, BioXtra, ~0.1 M in H2O
HCl
H0636
Hydrochloric acid solution, protein sequencing grade, liquid
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
71687
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
HNaO
72068
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
HNaO
S2567
Sodium hydroxide solution, volumetric, 1.0 M NaOH (1.0N)
HNaO
S2770
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
HNaO
609765
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
HNa16O