EMAIL THIS PAGE TO A FRIEND
Molecular Pharmacology
Rational Engineering Defines a Molecular Switch That Is Essential for Activity of Spider-Venom Peptides against the Analgesics Target NaV1.7.
PMID 26429937