EMAIL THIS PAGE TO A FRIEND
Mutation research
Protective effect of hydroxychavicol, a phenolic component of betel leaf, against the tobacco-specific carcinogens.
PMID 2643023
Abstract
The phenolic compound, hydroxychavicol (HC), present in betel leaf, was synthesised and tested for its antimutagenic effect against the mutagenicity of the 2 tobacco-specific N-nitrosamines (TSNA), N'-nitrosonornicotine (NNN) and 4-(nitrosomethylamino)-1-(3-pyridyl)-1-butanone (NNK), in 2 different test systems, viz. the Ames Salmonella/microsome assay and the micronucleus test using Swiss male mice. We are reporting the synthesis of HC of a high degree of purity. We observed that HC suppressed the mutagenic effects of NNN and NNK in both test systems used. These results indicate that HC may have a role to play in reducing the risk of oral cancer in betel quid with tobacco chewers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
18078
4-Allylpyrocatechol, analytical standard
C9H10O2