EMAIL THIS PAGE TO A FRIEND
Steroids
Microwave-assisted extraction and a new determination method for total steroid saponins from Dioscorea zingiberensis C.H. Wright.
PMID 26432686
Abstract
An efficient microwave-assisted extraction (MAE) technique was applied to isolate total steroid saponins from Dioscorea zingiberensis C.H. Wright (DZW). The optimal extracting conditions were established as 75% ethanol as solvent, ratio of solid/liquid 1:20 (g/ml), temperature 75 °C, irradiation power 600 W and three extraction cycles of 6 min each. Scanning electron microscopy (SEM) images of DZW processed by four different extractions provided visual evidence of the disruption effect on DZW. Diosgenin was quantified by HPLC and examined further by LC-ESI/MS after acid hydrolysis. Total steroid saponins were calculated using diosgenin from total steroid saponins. The MAE procedure was optimized, validated and compared with other conventional extraction processes. This report provides a convenient technology for the extraction and quantification of total saponins of DZW combining MAE with HPLC and LC-ESI/MS for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1634
Diosgenin, ≥93%
C27H42O3