EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Heart and Circulatory Physiology
N-acetylcysteine reverses diastolic dysfunction and hypertrophy in familial hypertrophic cardiomyopathy.
PMID 26432840