EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Lung Cellular and Molecular Physiology
Imatinib attenuates inflammation and vascular leak in a clinically relevant two-hit model of acute lung injury.
PMID 26432864