EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Graphene oxide/polyethyleneglycol composite coated stir bar for sorptive extraction of fluoroquinolones from chicken muscle and liver.
PMID 26433263
Abstract
Graphene oxide (GO) is an ideal adsorbent for polar and less polar compounds due to its hexagonal carbon network structure with oxygen-containing groups, while its strong hydrophilicity and water solubility limited its application in sample pretreatment techniques. Herein, GO was composited with polyethyleneglycol (PEG) or polyaniline (PAN) through intermolecular interactions to improve its stability, and the GO/PEG and GO/PAN composite coated stir bars were prepared by sol-gel technique. Compared with GO/PAN composite and polydimethylsiloxane (PDMS) coated stir bar, the prepared GO/PEG composite coated stir bar exhibited higher extraction efficiency for five fluoroquinolones (FQs). Based on it, a method of GO/PEG composite coated stir bar sorptive extraction (SBSE) combined with high-performance liquid chromatography-fluorescence detector (HPLC-FLD) was proposed. The factors influencing SBSE, such as sample pH, salt effect, stirring rate, extraction time, desorption solvent and desorption time, were optimized, and the analytical performance of the developed SBSE-HPLC-FLD method was evaluated. The limits of detection (LODs) for five FQs were in the range of 0.0045-0.0079μgL(-1), and the enrichment factors (EFs) were in the range of 41.5-65.5-fold (theoretical enrichment factor was 100-fold). The reproducibility was also investigated at concentrations of 0.05μgL(-1) and the relative standard deviations (RSDs, n=6) were found to be in the range of 4.6-12.1%. The proposed method was successfully applied for the determination of FQs in chicken muscle and chicken liver samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
242284
Aniline, ACS reagent, ≥99.5%
C6H7N
132934
Aniline, ReagentPlus®, 99%
C6H7N
270997
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
CH2Cl2
439223
Dichloromethane, for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
CH2Cl2
270563
Dichloromethane, ≥99.9%
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
07-2205
Dichloromethane, suitable for PCB analysis
CH2Cl2
07-2206
Dichloromethane, JIS 5000, for residue analysis
CH2Cl2
07-2230
Dichloromethane, for HPLC
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2
723851
Dichloromethane solution, contains 10 % (v/v) methanol
CH2Cl2
L5626
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
C6H14N2O2 · HCl
L8662
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H14N2O2 · HCl
L7039
L-Lysine monohydrochloride, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C6H14N2O2 · HCl
62929
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
C6H14N2O2 · HCl
D4551
N,N-Dimethylformamide, for molecular biology, ≥99%
C3H7NO
227056
N,N-Dimethylformamide, anhydrous, 99.8%
C3H7NO
648531
N,N-Dimethylformamide, for HPLC, ≥99.9%
C3H7NO
437573
N,N-Dimethylformamide, ACS reagent, ≥99.8%
C3H7NO
379824
Potassium persulfate, 99.99% trace metals basis
K2O8S2
216224
Potassium persulfate, ACS reagent, ≥99.0%
K2O8S2
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S3139
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P
S2554
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
H2NaO4P
229903
Sodium phosphate monobasic, 99.999% trace metals basis
H2NaO4P
S0751
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
H2NaO4P
74092
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
H2NaO4P