EMAIL THIS PAGE TO A FRIEND
European Journal of Medicinal Chemistry
Synthesis and pharmacological evaluation of some new fluorine containing hydroxypyrazolines as potential anticancer and antioxidant agents.
PMID 26433616