EMAIL THIS PAGE TO A FRIEND
Nature Communications
Measuring fast stochastic displacements of bio-membranes with dynamic optical displacement spectroscopy.
PMID 26437911
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
RES6003H-B7
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
H7523
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
C8H18N2O4S
H6147
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
C8H18N2O4S
54457
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
C8H18N2O4S
83264
HEPES buffer solution, 1 M in H2O
C8H18N2O4S
P1223
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
C41H78NO8P
P5655
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
H2KO4P
RES20760-A7
Potassium phosphate monobasic, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H2KO4P
P8416
Potassium phosphate monobasic, plant cell culture tested, ≥99%
H2KO4P
P9791
Potassium phosphate monobasic, for molecular biology, ≥98.0%
H2KO4P
229806
Potassium phosphate monobasic, 99.99% trace metals basis
H2KO4P
P5379
Potassium phosphate monobasic, ReagentPlus®
H2KO4P
60218
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
H2KO4P
60220
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
H2KO4P
04243
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
H2KO4P