EMAIL THIS PAGE TO A FRIEND
Dental Materials Journal
The influence of surface roughness and surface dynamics on the attachment of Methicillin-Resistant Staphylococcus aureus onto orthodontic retainer materials.
PMID 26438981
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442679
Hexadecane, analytical standard
C16H34