EMAIL THIS PAGE TO A FRIEND
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy
Validated derivative and ratio derivative spectrophotometric methods for the simultaneous determination of levocetirizine dihydrochloride and ambroxol hydrochloride in pharmaceutical dosage form.
PMID 26439526