EMAIL THIS PAGE TO A FRIEND
Cell Biochemistry and Function
Esculetin induces apoptosis in human colon cancer cells by inducing endoplasmic reticulum stress.
PMID 26439795
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246573
6,7-Dihydroxycoumarin, 98%
C9H6O4