EMAIL THIS PAGE TO A FRIEND
Journal of Fluorescence
Determination of Some Non-sedating Antihistamines via Their Native Fluorescence and Derivation of Some Quantitative Fluorescence Intensity - Structure Relationships.
PMID 26439930