EMAIL THIS PAGE TO A FRIEND
Oncotarget
The timing of surgery after neoadjuvant radiotherapy influences tumor dissemination in a preclinical model.
PMID 26440148
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1165
Pimonidazole, ≥98% (HPLC)
C11H18N4O3