EMAIL THIS PAGE TO A FRIEND
Journal of animal science
Effects of alternate day feeding of dried distiller's grains plus solubles in forage-fed steers on intake, ruminal fermentation and passage rates, and serum nonesterified fatty acid.
PMID 26440176
Abstract
Four ruminally and duodenally cannulated Holstein steers (BW = 449 ± 7.3 kg) were used to examine the effects of feeding either dried distiller's grains plus solubles (DG) or grass hay on alternate days (every other day) on intake, ruminal fermentation and passage rates, and serum NEFA in forage-fed steers. Steers were assigned to 1 of 4 dietary treatments in a 4 × 4 Latin square: 1) only hay (CON), 2) hay and 0.4% of BW as DG DM daily (DG7), 3) hay daily and 0.8% BW DG every other day (DG2), and 4) alternate day feeding of hay and 0.8% of BW as DG (DGA). Treatment periods consisted of 13 d of adaptation and 8 d of collecting digesta and blood. Over the entire collection period, DMI was decreased ( = 0.004) for DGA compared with other treatments (13.0 ± 0.8, 12.7 ± 0.8, 13.3 ± 0.8, and 10.9 ± 0.8 kg/d for CON, DG7, DG2, and DGA, respectively). Immediately after feeding on days supplement was fed to DG2 and DGA (supplemented days [SUP]), ruminal pH of DGA was less than other treatments but by the end of the day was greater than other treatments (treatment × time, < 0.001). At feeding time on nonsupplemented days (NSUP), ruminal pH of DGA steers was greater than other treatments but was similar (treatment × time, < 0.001) to DG2 and CON by 5 h after feeding. Total concentrations of VFA were similar ( = 0.09) among treatments on SUP; however, on NSUP, total VFA concentrations were least in DGA from feeding until 4 h after feeding (treatment × time, = 0.02). No differences ( ≥ 0.06) were observed among treatments for apparent ruminal, total intestinal, and total tract DM, OM, or CP digestibility. There were no differences ( = 0.36) in serum NEFA among treatments on SUP; however, on NSUP, steers fed DGA (209.5 ± 12.7 m) had greater ( < 0.01) NEFA compared with other treatments (84.4 ± 12.7, 88.0 ± 12.7, and 77.7 ± 12.7 m for CON, DG7, and DG2, respectively). The DGA feeding strategy influenced DMI and ruminal kinetics and circulating NEFA without impacting total tract digestibility.