EMAIL THIS PAGE TO A FRIEND
Toxicology and Applied Pharmacology
The effects of gestational and chronic atrazine exposure on motor behaviors and striatal dopamine in male Sprague-Dawley rats.
PMID 26440580