EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
Omeprazole induces NAD(P)H quinone oxidoreductase 1 via aryl hydrocarbon receptor-independent mechanisms: Role of the transcription factor nuclear factor erythroid 2-related factor 2.
PMID 26441083