EMAIL THIS PAGE TO A FRIEND
Chirality
Chiral Pool-Based Synthesis of Naphtho-Fused Isocoumarins.
PMID 26441372
Abstract
A variety of chiral derivatives of benzo[d]naphtho[1,2-b]pyran-6-one were prepared in a single step by Et3 N-mediated condensation of homophthalic anhydride with different derivatives of (S)-amino acid chlorides at -5 °C by employing a chiral pool methodology.