EMAIL THIS PAGE TO A FRIEND
Frontiers in Physiology
Caloric restriction induces energy-sparing alterations in skeletal muscle contraction, fiber composition and local thyroid hormone metabolism that persist during catch-up fat upon refeeding.
PMID 26441673
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB1572
Anti-Parvalbumin Antibody, ascites fluid, clone PARV-19, Chemicon®