EMAIL THIS PAGE TO A FRIEND
European Journal of Obstetrics, Gynecology, and Reproductive Biology
Mouse preantral follicle development in two-dimensional and three-dimensional culture systems after ovarian tissue vitrification.
PMID 26444332