EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
A novel UHPLC method for the rapid and simultaneous determination of daidzein, genistein and equol in human urine.
PMID 26444491
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H20202
4-Hydroxybenzophenone, 98%
C13H10O2
D7802
Daidzein, ≥98%, synthetic
C15H10O4