EMAIL THIS PAGE TO A FRIEND
Nature communications
An LSC epigenetic signature is largely mutation independent and implicates the HOXA cluster in AML pathogenesis.
PMID 26444494
Abstract
Acute myeloid leukaemia (AML) is characterized by subpopulations of leukaemia stem cells (LSCs) that are defined by their ability to engraft in immunodeficient mice. Here we show an LSC DNA methylation signature, derived from xenografts and integration with gene expression that is comprised of 71 genes and identifies a key role for the HOXA cluster. Most of the genes are epigenetically regulated independently of underlying mutations, although several are downstream targets of epigenetic modifier genes mutated in AML. The LSC epigenetic signature is associated with poor prognosis independent of known risk factors such as age and cytogenetics. Analysis of early haematopoietic progenitors from normal individuals reveals two distinct clusters of AML LSC resembling either lymphoid-primed multipotent progenitors or granulocyte/macrophage progenitors. These results provide evidence for DNA methylation variation between AML LSCs and their blast progeny, and identify epigenetically distinct subgroups of AML likely reflecting the cell of origin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
307823
Sodium hydrogen sulfate, technical grade
HNaO4S