EMAIL THIS PAGE TO A FRIEND
Clinical Hemorheology and Microcirculation
Human microvascular endothelial cells displaying reduced angiogenesis and increased uptake of lipids during in vitro culture.
PMID 26444614