EMAIL THIS PAGE TO A FRIEND
Journal of Alzheimer's disease : JAD
UCH-L1 Inhibition Decreases the Microtubule-Binding Function of Tau Protein.
PMID 26444754
Abstract
Ubiquitin C-terminal hydrolase L1 (UCH-L1) is critical for protein degradation and free ubiquitin recycling. In Alzheimer's disease brains, UCH-L1 is negatively related to neurofibrillary tangles whose major component is hyperphosphorylated tau protein, but the direct action of UCH-L1 on tau has not been reported. In the current study, mouse neuroblastoma Neuro2a (N2a) cells were treated by the different concentrations of UCH-L1 inhibitor LDN (2.5, 5 and 10 μM) to inhibit the hydrolase activity of UCH-L1. In addition, we also used UCH-L1 siRNA to treat the HEK293/tau441 cells to decrease the expression of UCH-L1. After LDN and UCH-L1 siRNA treatment, we used immunofluorescence, immunoprecipitation, and tau-microtubule binding assay to measure the microtubule-binding ability and post-translational modifications of tau protein. All the results presented that both inhibition of the activity and expression of UCH-L1 induced the decreased microtubule-binding ability and increased phosphorylation of tau protein. Abnormal aggregation and ubiquitination of tau protein was also observed after UCH-L1 inhibition. The above results suggested that aggregation of tau protein might be devoted to the abnormal post-translational modifications of tau protein. Our study first indicates that dysfunction of UCH-L1 most likely affected normal biological function of tau protein through decreasing degradation of ubiquitinated and hyperphosphorylated tau.