EMAIL THIS PAGE TO A FRIEND
Journal of Alzheimer'S Disease
Chronic Hypoxia-Induced Autophagy Aggravates the Neuropathology of Alzheimer's Disease through AMPK-mTOR Signaling in the APPSwe/PS1dE9 Mouse Model.
PMID 26444766
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D161403
5,5-Dimethylhydantoin, 97%
C5H8N2O2