EMAIL THIS PAGE TO A FRIEND
Physiological Research
Nano-colloid printing of functionalized PLA-b-PEO copolymers: tailoring the surface pattern of adhesive motif and its effect on cell attachment.
PMID 26447596