EMAIL THIS PAGE TO A FRIEND
International Journal of Impotence Research
Additive effects of Artemisia capillaris extract and scopoletin on the relaxation of penile corpus cavernosum smooth muscle.
PMID 26447600