EMAIL THIS PAGE TO A FRIEND
Aquatic Toxicology (Amsterdam, Netherlands)
n-TiO2 and CdCl2 co-exposure to titanium dioxide nanoparticles and cadmium: Genomic, DNA and chromosomal damage evaluation in the marine fish European sea bass (Dicentrarchus labrax).
PMID 26448269