EMAIL THIS PAGE TO A FRIEND
Journal of proteome research
Mass Spectrometric N-Glycan Analysis of Haptoglobin from Patient Serum Samples Using a 96-Well Plate Format.
PMID 26448449
Abstract
Alterations in glycosylation of serum glycoproteins can provide unique and highly specific fingerprints of malignancy. Our previous mass spectrometric study revealed that the bifucosylation level of serum haptoglobin was distinctly increased in hepatocellular carcinoma (HCC) patients versus liver cirrhosis of all three major etiologies. We have thus developed a method for the analysis of large numbers of serum samples based on a 96-well plate platform for the evaluation of fucosylation changes of serum haptoglobin between HCC versus cirrhosis. Haptoglobin was isolated from the serum of individual patient samples based on an HPLC column immobilized with antihaptoglobin antibody via hydrazide immobilization chemistry. Only 10 μL of serum was required for glycan extraction and processing for MALDI-QIT mass spectrometry analysis using the 96-well plate format. The bifucosylation degrees of haptoglobin in individuals were calculated using a quantitative glycomics method. The MS data confirmed that the bifucosylated tetra-anntenary glycan was upregulated in HCC samples of all etiologies. This study provides a parallel method for processing glycan content for haptoglobin and evaluating detailed changes in glycan structures for a potentially large cohort of clinical serum samples.