EMAIL THIS PAGE TO A FRIEND
Veterinary and Comparative Orthopaedics and Traumatology : V.C.O.T
Influence of a titanium mesh on the management of segmental long bone defects. An experimental study in a canine ulnar model.
PMID 26449275
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1019
(±)-Thiopental
C11H18N2O2S