EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
The tumor vessel targeting agent NGR-TNF controls the different stages of the tumorigenic process in transgenic mice by distinct mechanisms.
PMID 26451306
Abstract
NGR-TNF is a vascular targeting agent in advanced clinical development, coupling tumor necrosis factor-α (TNF) with the CNGRCG peptide, which targets a CD13 isoform specifically expressed by angiogenic vessels. Antitumor efficacy of NGR-TNF has been described in different transplantation tumor models. Nevertheless, the mechanism underlying its activity is not fully understood. In the wild type and in the immunodeficient (RAG