Chemical research in toxicology
Polychlorinated Biphenyl Quinone Metabolite Promotes p53-Dependent DNA Damage Checkpoint Activation, S-Phase Cycle Arrest and Extrinsic Apoptosis in Human Liver Hepatocellular Carcinoma HepG2 Cells.
PMID 26451628
Polychlorinated biphenyls (PCBs) are a group of persistent organic pollutants. The toxic behavior and mechanism of PCBs individuals and congeners have been extensively investigated. However, there is only limited information on their metabolites. Our previous studies have shown that a synthetic PCB metabolite, PCB29-pQ, causes oxidative damage with the evidence of cytotoxicity, genotoxicity, and mitochondrial-derived intrinsic apoptosis. Here, we investigate the effects of PCB29-pQ on DNA damage checkpoint activation, cell cycle arrest, and death receptor-related extrinsic apoptosis in human liver hepatocellular carcinoma HepG2 cells. Our results illustrate that PCB29-pQ increases the S-phase cell population by down-regulating cyclins A/D1/E, cyclin-dependent kinases (CDK 2/4/6), and cell division cycle 25A (CDC25A) and up-regulating p21/p27 protein expressions. PCB29-pQ also induces apoptosis via the up-regulation of Fas/FasL and the activation of caspase 8/3. Moreover, p53 plays a pivotal role in PCB29-pQ-induced cell cycle arrest and apoptosis via the activation of ATM/Chk2 and ATR/Chk1 checkpoints. Cell cycle arrest and apoptotic cell death were attenuated by the pretreatment with antioxidant N-acetyl-cysteine (NAC). Taken together, these results demonstrate that PCB29-pQ induces oxidative stress and promotes p53-dependent DNA damage checkpoint activation, S-phase cycle arrest, and extrinsic apoptosis in HepG2 cells.
Related Materials
Product #
Molecular Formula
Add to Cart
N-Acetyl-Asp-Glu-Val-Asp-al, ≥95%, powder