EMAIL THIS PAGE TO A FRIEND
Journal of Food Science
Molecular Structural Characteristics of Polysaccharide Fractions from Canarium album (Lour.) Raeusch and Their Antioxidant Activities.
PMID 26452687
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471321
2-(Diethylamino)ethanol, ≥99.5%
C6H15NO